The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256295.1 | NP_001243224.1 | 10 - 11 | 1655 | 92 |
NM_005239.5 | NP_005230.1 | 9 - 10 | 1485 | 92 | |
XM_005260935.1 | XP_005260992.1 | 9 - 10 | 1558 | 92 | |
XM_017028290.1 | XP_016883779.1 | 9 - 10 | 4155 | 92 | |
GenBank mRNA | AK315563.1 | - | 9 - 10 | 1388 | 92 |
BC017040.1 | - | 9 - 10 | 1357 | 92 | |
BC042954.2 | - | 9 - 10 | 1387 | 92 | |
BT006838.1 | - | 8 - 9 | 1195 | 92 | |
J04102.1 | - | 9 - 10 | 1486 | 92 | |
M11922.1 | - | 2 - 3 | 226 | 92 |