The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002021.2 | NP_001002021.2 | 11 - 12 | 1394 | 54 |
NM_002626.5 | NP_002617.3 | 8 - 9 | 916 | 54 | |
XM_005261135.3 | XP_005261192.1 | 9 - 10 | 979 | 54 | |
XM_005261137.4 | XP_005261194.1 | 8 - 9 | 1007 | 54 | |
XM_006724011.1 | XP_006724074.1 | 12 - 13 | 1409 | 54 | |
XM_006724012.1 | XP_006724075.1 | 9 - 10 | 1125 | 54 | |
XM_011529603.2 | XP_011527905.1 | 2 - 3 | 578 | 54 | |
XM_017028368.1 | XP_016883857.1 | 11 - 12 | 1373 | 54 | |
XM_017028369.1 | XP_016883858.1 | 10 - 11 | 1144 | 54 | |
XM_017028370.1 | XP_016883859.1 | 10 - 11 | 1272 | 54 | |
XM_017028371.1 | XP_016883860.1 | 9 - 10 | 1050 | 54 | |
XM_017028372.1 | XP_016883861.1 | 9 - 10 | 1096 | 54 | |
GenBank mRNA | AB209127.1 | - | 7 - 8 | 3076 | 54 |
AK091488.1 | - | 9 - 10 | 808 | 54 | |
AK303605.1 | - | 9 - 10 | 1121 | 54 | |
AK308437.1 | - | 8 - 9 | 877 | 54 | |
BC006422.1 | - | 11 - 12 | 1340 | 54 | |
BC007536.1 | - | 8 - 9 | 859 | 54 | |
BC008964.2 | - | 8 - 9 | 864 | 54 | |
BC009919.2 | - | 8 - 9 | 864 | 54 | |
BC018295.1 | - | 2 - 3 | 210 | 54 | |
BU177678.1 | - | 6 - 7 | 659 | 54 | |
BX537446.1 | - | 1 - 2 | 814 | 54 | |
X15573.1 | - | 8 - 9 | 899 | 54 |