The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004327.3 | NP_004318.3 | 18 - 19 | 3774 | 83 |
NM_021574.2 | NP_067585.2 | 17 - 18 | 3642 | 83 | |
GenBank mRNA | AB209991.1 | - | 18 - 19 | 3523 | 83 |
AF487522.1 | - | 3 - 4 | 235 | 83 | |
AK122842.1 | - | 1 - 2 | 956 | 83 | |
AK303241.1 | - | 18 - 19 | 2208 | 83 | |
BC066122.1 | - | 18 - 19 | 3605 | 83 | |
X02596.1 | - | 18 - 19 | 3666 | 83 | |
Y00661.1 | - | 17 - 18 | 3642 | 83 |