Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAK2
Entrez Gene ID
66008
Gene Name
trafficking kinesin protein 2
Gene Aliases
ALS2CR3, CALS-C, GRIF-1, GRIF1, MILT2, OIP98
Chromosome Location
Chr.2: 201377207 - 201451596 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015049.2 NP_055864.2 1 - 2 246 76
XM_017004772.1 XP_016860261.1 1 - 2 227 76
GenBank mRNA AB038951.1 - 1 - 2 181 76
AK054763.1 - 1 - 2 150 76
AK314384.1 - 1 - 2 105 76
AL832573.2 - 1 - 2 262 76
BC040668.2 - 1 - 2 197 76
BC041330.2 - 1 - 2 197 76
BC048093.1 - 1 - 2 197 76
DA414989.1 - 1 - 2 246 76

View Full Product Details