The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004521.2 | NP_004512.1 | 10 - 11 | 1428 | 110 |
XM_017016224.1 | XP_016871713.1 | 10 - 11 | 1034 | 110 | |
GenBank mRNA | AK289413.1 | - | 10 - 11 | 1374 | 110 |
AK307502.1 | - | 11 - 12 | 1125 | 110 | |
BC010885.1 | - | 10 - 11 | 1364 | 110 | |
BC065267.1 | - | 10 - 11 | 1385 | 110 | |
BC080604.1 | - | 10 - 11 | 1359 | 110 | |
BC111042.1 | - | 10 - 11 | 1368 | 110 | |
BC126279.1 | - | 10 - 11 | 1181 | 110 | |
BC126281.1 | - | 10 - 11 | 1181 | 110 | |
FJ224324.1 | - | 10 - 11 | 1271 | 110 | |
X65873.1 | - | 10 - 11 | 1271 | 110 |