The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004521.2 | NP_004512.1 | 3 - 4 | 763 | 64 |
XM_017016224.1 | XP_016871713.1 | 3 - 4 | 369 | 64 | |
GenBank mRNA | AK289413.1 | - | 3 - 4 | 709 | 64 |
AK307502.1 | - | 4 - 5 | 460 | 64 | |
BC010885.1 | - | 3 - 4 | 699 | 64 | |
BC065267.1 | - | 3 - 4 | 720 | 64 | |
BC080604.1 | - | 3 - 4 | 694 | 64 | |
BC111042.1 | - | 3 - 4 | 703 | 64 | |
BC126279.1 | - | 3 - 4 | 516 | 64 | |
BC126281.1 | - | 3 - 4 | 516 | 64 | |
FJ224324.1 | - | 3 - 4 | 606 | 64 | |
X65873.1 | - | 3 - 4 | 606 | 64 |