The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004521.2 | NP_004512.1 | 9 - 10 | 1287 | 152 |
XM_017016224.1 | XP_016871713.1 | 9 - 10 | 893 | 152 | |
GenBank mRNA | AK289413.1 | - | 9 - 10 | 1233 | 152 |
AK307502.1 | - | 10 - 11 | 984 | 152 | |
BC010885.1 | - | 9 - 10 | 1223 | 152 | |
BC065267.1 | - | 9 - 10 | 1244 | 152 | |
BC080604.1 | - | 9 - 10 | 1218 | 152 | |
BC111042.1 | - | 9 - 10 | 1227 | 152 | |
BC126279.1 | - | 9 - 10 | 1040 | 152 | |
BC126281.1 | - | 9 - 10 | 1040 | 152 | |
FJ224324.1 | - | 9 - 10 | 1130 | 152 | |
X65873.1 | - | 9 - 10 | 1130 | 152 |