Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPIN3
Entrez Gene ID
64900
Gene Name
lipin 3
Gene Aliases
LIPN3L, SMP2
Chromosome Location
Chr.20: 41340779 - 41360585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301860.1 NP_001288789.1 18 - 19 2466 89
NR_126051.1 18 - 19 2384 89
XM_011528996.1 XP_011527298.1 18 - 19 2632 89
XM_011528997.2 XP_011527299.1 19 - 20 2838 89
XM_011528998.2 XP_011527300.1 18 - 19 2655 89
XM_011528999.2 XP_011527301.1 19 - 20 2785 89
XM_011529000.1 XP_011527302.1 18 - 19 2629 89
XM_011529001.1 XP_011527303.1 17 - 18 2311 89
XM_011529002.2 XP_011527304.1 17 - 18 2434 89
XM_017028020.1 XP_016883509.1 18 - 19 2461 89
GenBank mRNA BC140806.1 - 18 - 19 2347 89
BX647082.1 - 15 - 16 3444 89

View Full Product Details