The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252675.1 | NP_001239604.1 | 2 - 3 | 513 | 79 |
NM_001252677.1 | NP_001239606.1 | 2 - 3 | 513 | 79 | |
NM_032501.3 | NP_115890.2 | 2 - 3 | 513 | 79 | |
XM_006723659.1 | XP_006723722.1 | 2 - 3 | 474 | 79 | |
XM_011529388.1 | XP_011527690.1 | 2 - 3 | 474 | 79 | |
GenBank mRNA | AK127566.1 | - | 2 - 3 | 459 | 79 |
AK314440.1 | - | 2 - 3 | 436 | 79 | |
AL832939.1 | - | 2 - 3 | 197 | 79 | |
BC039261.1 | - | 2 - 3 | 472 | 79 | |
BC044588.1 | - | 2 - 3 | 438 | 79 | |
BC055008.1 | - | 2 - 3 | 438 | 79 | |
BC114612.1 | - | 2 - 3 | 304 | 79 | |
BM150326.1 | - | 2 - 3 | 513 | 79 |