Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
Chromosome Location
Chr.20: 25006230 - 25058182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252675.1 NP_001239604.1 9 - 10 1522 97
NM_001252676.1 NP_001239605.1 8 - 9 1355 97
NM_001252677.1 NP_001239606.1 9 - 10 1528 97
NM_032501.3 NP_115890.2 9 - 10 1528 97
XM_006723659.1 XP_006723722.1 5 - 6 874 97
XM_011529388.1 XP_011527690.1 8 - 9 1313 97
GenBank mRNA AB058749.1 - 4 - 5 445 97
AK027817.1 - 3 - 4 351 97
AK092295.1 - 7 - 8 937 97
AK125058.1 - 7 - 8 2731 97
AK127566.1 - 9 - 10 1474 97
AK296306.1 - 8 - 9 1352 97
AK314440.1 - 9 - 10 1451 97
AL832939.1 - 5 - 6 597 97
BC032076.1 - 2 - 3 216 97
BC039261.1 - 9 - 10 1487 97
BC044588.1 - 9 - 10 1447 97
BC055008.1 - 9 - 10 1447 97

View Full Product Details