The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000059.3 | NP_000050.2 | 11 - 12 | 7062 | 177 |
GenBank mRNA | DQ897648.1 | - | 10 - 11 | 6835 | 177 |
U43746.1 | - | 11 - 12 | 7063 | 177 |