Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRCA2
Entrez Gene ID
675
Gene Name
BRCA2, DNA repair associated
Gene Aliases
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11
Chromosome Location
Chr.13: 32315480 - 32399672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000059.3 NP_000050.2 15 - 16 7846 118
GenBank mRNA DQ897648.1 - 14 - 15 7619 118
U43746.1 - 15 - 16 7847 118

View Full Product Details