Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP2M1
Entrez Gene ID
1173
Gene Name
adaptor related protein complex 2 mu 1 subunit
Gene Aliases
AP50, CLAPM1, mu2
Chromosome Location
Chr.3: 184174846 - 184184091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025205.1 NP_001020376.1 2 - 3 227 109
NM_004068.3 NP_004059.2 2 - 3 227 109
GenBank mRNA AK057883.1 - 2 - 3 218 109
AK304510.1 - 2 - 3 198 109
BC004996.1 - 2 - 3 205 109
BC013796.1 - 2 - 3 145 109
BC014030.1 - 2 - 3 195 109
BI461415.1 - 2 - 3 255 109
D63475.1 - 2 - 3 156 109
DB261680.1 - 2 - 3 227 109
U36188.1 - 1 - 2 116 109

View Full Product Details