Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDK4
Entrez Gene ID
5166
Gene Name
pyruvate dehydrogenase kinase 4
Gene Aliases
-
Chromosome Location
Chr.7: 95583497 - 95596613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002612.3 NP_002603.1 10 - 11 1420 153
GenBank mRNA AK096428.1 - 11 - 12 1219 153
AK098151.1 - 6 - 7 734 153
AK223328.1 - 10 - 11 1316 153
AK312424.1 - 10 - 11 1326 153
BC040239.1 - 10 - 11 1300 153
U54617.1 - 10 - 11 1322 153

View Full Product Details