The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002612.3 | NP_002603.1 | 5 - 6 | 933 | 80 |
GenBank mRNA | AK096428.1 | - | 6 - 7 | 732 | 80 |
AK098151.1 | - | 1 - 2 | 247 | 80 | |
AK223328.1 | - | 5 - 6 | 829 | 80 | |
AK312424.1 | - | 5 - 6 | 839 | 80 | |
BC040239.1 | - | 5 - 6 | 813 | 80 | |
U54617.1 | - | 5 - 6 | 835 | 80 |