The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102469.1 | NP_001095939.1 | 5 - 6 | 669 | 132 |
XM_005270049.3 | XP_005270106.1 | 7 - 8 | 961 | 132 | |
XM_011540083.2 | XP_011538385.1 | 7 - 8 | 998 | 132 | |
XM_011540084.2 | XP_011538386.1 | 6 - 7 | 883 | 132 | |
XM_011540085.2 | XP_011538387.1 | 7 - 8 | 998 | 132 | |
XM_011540086.2 | XP_011538388.1 | 7 - 8 | 998 | 132 | |
GenBank mRNA | EF426483.1 | - | 5 - 6 | 669 | 132 |