The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322364.1 | NP_001309293.1 | 2 - 3 | 210 | 121 |
NM_033497.2 | NP_277032.1 | 3 - 4 | 332 | 121 | |
NM_033498.2 | NP_277033.1 | 3 - 4 | 386 | 121 | |
NM_033500.2 | NP_277035.2 | 3 - 4 | 386 | 214 | |
XM_011539732.1 | XP_011538034.1 | 3 - 4 | 293 | 214 | |
GenBank mRNA | AF073786.2 | - | 2 - 3 | 251 | 214 |
DB088955.1 | - | 3 - 4 | 336 | 121 |