The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005245622.3 | XP_005245679.1 | 16 - 17 | 4057 | 123 |
XM_011510166.2 | XP_011508468.1 | 18 - 19 | 4484 | 123 | |
XM_017002849.1 | XP_016858338.1 | 18 - 19 | 7090 | 123 | |
XM_017002851.1 | XP_016858340.1 | 18 - 19 | 7068 | 123 | |
XM_017002853.1 | XP_016858342.1 | 18 - 19 | 4528 | 123 | |
XM_017002854.1 | XP_016858343.1 | 16 - 17 | 4060 | 123 | |
XM_017002855.1 | XP_016858344.1 | 16 - 17 | 4038 | 123 | |
GenBank mRNA | AK022662.1 | - | 8 - 9 | 1927 | 123 |