The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015247.2 | NP_056062.1 | 2 - 3 | 149 | 150 |
XM_005255812.2 | XP_005255869.1 | 2 - 3 | 109 | 150 | |
XM_006721149.1 | XP_006721212.1 | 2 - 3 | 173 | 150 | |
XM_017022978.1 | XP_016878467.1 | 1 - 2 | 76 | 150 | |
GenBank mRNA | AJ250014.1 | - | 2 - 3 | 125 | 150 |
AK225202.1 | - | 2 - 3 | 129 | 150 | |
DB119480.1 | - | 2 - 3 | 172 | 150 |