The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077440.1 | NP_001070908.1 | 4 - 5 | 1269 | 83 |
NM_001301038.1 | NP_001287967.1 | 4 - 5 | 1269 | 83 | |
NM_014739.2 | NP_055554.1 | 4 - 5 | 1275 | 83 | |
XM_005267237.3 | XP_005267294.1 | 4 - 5 | 1211 | 83 | |
XM_017011532.1 | XP_016867021.1 | 4 - 5 | 1211 | 83 | |
GenBank mRNA | AF249273.1 | - | 2 - 3 | 1016 | 83 |
BC016682.1 | - | 4 - 5 | 1214 | 83 | |
BC047887.1 | - | 4 - 5 | 1244 | 83 | |
BC056894.1 | - | 4 - 5 | 1256 | 83 | |
BC063846.1 | - | 4 - 5 | 1182 | 83 | |
BC078160.1 | - | 4 - 5 | 1181 | 83 | |
BC132780.1 | - | 4 - 5 | 1219 | 83 | |
BC144281.1 | - | 4 - 5 | 1213 | 83 | |
BC144299.1 | - | 4 - 5 | 1219 | 83 | |
D79986.1 | - | 4 - 5 | 1275 | 83 |