The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001376.4 | NP_001367.2 | 34 - 35 | 7183 | 62 |
GenBank mRNA | AB002323.3 | - | 34 - 35 | 7055 | 62 |
AB290157.1 | - | 34 - 35 | 7183 | 62 | |
AK095415.1 | - | 4 - 5 | 796 | 62 | |
AL833600.1 | - | 6 - 7 | 1181 | 62 |