Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIKFYVE
Entrez Gene ID
200576
Gene Name
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location
Chr.2: 208266178 - 208358751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015040.3 NP_055855.2 15 - 16 2166 73
XM_011510778.2 XP_011509080.1 16 - 17 2216 73
XM_011510779.2 XP_011509081.1 16 - 17 2149 73
XM_011510780.2 XP_011509082.1 16 - 17 2234 73
XM_011510781.2 XP_011509083.1 16 - 17 2157 73
XM_011510782.2 XP_011509084.1 16 - 17 2216 73
XM_011510783.2 XP_011509085.1 15 - 16 2048 73
XM_011510784.2 XP_011509086.1 15 - 16 2066 73
XM_011510785.2 XP_011509087.1 15 - 16 1988 73
XM_011510786.2 XP_011509088.1 14 - 15 1883 73
XM_011510787.1 XP_011509089.1 14 - 15 1892 73
XM_011510788.1 XP_011509090.1 13 - 14 1875 73
XM_011510789.2 XP_011509091.1 12 - 13 1697 73
XM_011510790.1 XP_011509092.1 14 - 15 1994 73
XM_011510792.2 XP_011509094.1 16 - 17 2216 73
XM_017003568.1 XP_016859057.1 15 - 16 2052 73
XM_017003569.1 XP_016859058.1 14 - 15 1884 73
XM_017003570.1 XP_016859059.1 12 - 13 1733 73
XM_017003571.1 XP_016859060.1 11 - 12 1529 73
XM_017003572.1 XP_016859061.1 14 - 15 1870 73
XM_017003573.1 XP_016859062.1 11 - 12 1578 73
XM_017003574.1 XP_016859063.1 10 - 11 1249 73
GenBank mRNA AK127769.1 - 14 - 15 1903 73
AY457063.1 - 15 - 16 2170 73
BC017736.2 - 14 - 15 1994 73
EU794609.1 - 15 - 16 2170 73

View Full Product Details