Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIKFYVE
Entrez Gene ID
200576
Gene Name
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location
Chr.2: 208266178 - 208358751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015040.3 NP_055855.2 35 - 36 5527 142
XM_011510778.2 XP_011509080.1 36 - 37 5577 142
XM_011510779.2 XP_011509081.1 36 - 37 5510 142
XM_011510780.2 XP_011509082.1 36 - 37 5595 142
XM_011510781.2 XP_011509083.1 36 - 37 5518 142
XM_011510783.2 XP_011509085.1 35 - 36 5409 142
XM_011510784.2 XP_011509086.1 35 - 36 5427 142
XM_011510785.2 XP_011509087.1 35 - 36 5349 142
XM_011510786.2 XP_011509088.1 34 - 35 5244 142
XM_011510787.1 XP_011509089.1 34 - 35 5253 142
XM_011510788.1 XP_011509090.1 33 - 34 5236 142
XM_011510789.2 XP_011509091.1 32 - 33 5058 142
XM_011510790.1 XP_011509092.1 34 - 35 5355 142
XM_017003568.1 XP_016859057.1 35 - 36 5413 142
XM_017003569.1 XP_016859058.1 34 - 35 5245 142
XM_017003570.1 XP_016859059.1 32 - 33 5094 142
XM_017003571.1 XP_016859060.1 31 - 32 4890 142
XM_017003572.1 XP_016859061.1 34 - 35 5231 142
XM_017003573.1 XP_016859062.1 31 - 32 4939 142
XM_017003574.1 XP_016859063.1 30 - 31 4610 142
GenBank mRNA AB023198.1 - 8 - 9 810 142
AK091482.1 - 16 - 17 2263 142
AY457063.1 - 35 - 36 5531 142
EU794609.1 - 35 - 36 5531 142

View Full Product Details