The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152671.3 | NP_689884.1 | 3 - 4 | 482 | 118 |
XM_011510788.1 | XP_011509090.1 | 3 - 4 | 482 | 118 | |
XM_017003570.1 | XP_016859059.1 | 3 - 4 | 508 | 118 | |
GenBank mRNA | BC032389.1 | - | 3 - 4 | 482 | 118 |