The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001671.3 | NP_001001671.3 | 25 - 26 | 3561 | 95 |
XM_011545507.2 | XP_011543809.2 | 26 - 27 | 3788 | 95 | |
XM_011545508.2 | XP_011543810.2 | 26 - 27 | 3701 | 95 | |
XM_011545510.2 | XP_011543812.1 | 19 - 20 | 4301 | 95 | |
XM_011545511.2 | XP_011543813.1 | 16 - 17 | 2812 | 95 | |
GenBank mRNA | AK122632.1 | - | 4 - 5 | 979 | 95 |
AK131412.1 | - | 22 - 23 | 2889 | 95 | |
AK131477.1 | - | 26 - 27 | 3621 | 95 |