The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001671.3 | NP_001001671.3 | 8 - 9 | 1282 | 81 |
XM_011545507.2 | XP_011543809.2 | 9 - 10 | 1509 | 81 | |
XM_011545508.2 | XP_011543810.2 | 9 - 10 | 1509 | 81 | |
XM_011545510.2 | XP_011543812.1 | 2 - 3 | 2022 | 81 | |
XM_017029511.1 | XP_016885000.1 | 9 - 10 | 1511 | 81 | |
GenBank mRNA | AK131412.1 | - | 4 - 5 | 537 | 81 |
AK131477.1 | - | 7 - 8 | 1171 | 81 | |
CN293540.1 | - | 2 - 3 | 294 | 81 |