The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100875.2 | NP_001094345.1 | 5 - 6 | 770 | 90 |
NM_001288779.1 | NP_001275708.1 | 6 - 7 | 965 | 90 | |
NM_001288780.1 | NP_001275709.1 | 6 - 7 | 996 | 90 | |
NM_152598.3 | NP_689811.2 | 5 - 6 | 851 | 90 | |
XM_005257095.2 | XP_005257152.2 | 6 - 7 | 927 | 90 | |
XM_005257096.2 | XP_005257153.2 | 6 - 7 | 872 | 90 | |
XM_005257097.3 | XP_005257154.2 | 7 - 8 | 1019 | 90 | |
XM_005257098.2 | XP_005257155.2 | 6 - 7 | 924 | 90 | |
XM_005257100.2 | XP_005257157.2 | 5 - 6 | 813 | 90 | |
XM_005257102.2 | XP_005257159.2 | 5 - 6 | 810 | 90 | |
XM_005257103.1 | XP_005257160.1 | 5 - 6 | 810 | 90 | |
XM_005257106.4 | XP_005257163.2 | 5 - 6 | 578 | 90 | |
XM_011524435.1 | XP_011522737.1 | 6 - 7 | 924 | 90 | |
XM_011524436.1 | XP_011522738.1 | 5 - 6 | 758 | 90 | |
XM_011524437.1 | XP_011522739.1 | 6 - 7 | 902 | 90 | |
GenBank mRNA | AK093076.1 | - | 5 - 6 | 804 | 90 |
AK122947.1 | - | 6 - 7 | 905 | 90 | |
BC035021.1 | - | 5 - 6 | 774 | 90 | |
BX648705.1 | - | 6 - 7 | 928 | 90 |