The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005923.3 | NP_005914.1 | 19 - 20 | 3037 | 64 |
XM_011535839.2 | XP_011534141.2 | 20 - 21 | 3361 | 64 | |
XM_017010870.1 | XP_016866359.1 | 20 - 21 | 3360 | 64 | |
XM_017010871.1 | XP_016866360.1 | 20 - 21 | 3360 | 64 | |
XM_017010872.1 | XP_016866361.1 | 20 - 21 | 3359 | 64 | |
XM_017010873.1 | XP_016866362.1 | 20 - 21 | 3358 | 64 | |
XM_017010874.1 | XP_016866363.1 | 19 - 20 | 3252 | 64 | |
XM_017010875.1 | XP_016866364.1 | 19 - 20 | 3251 | 64 | |
XM_017010876.1 | XP_016866365.1 | 18 - 19 | 2368 | 64 | |
XM_017010877.1 | XP_016866366.1 | 18 - 19 | 2605 | 64 | |
GenBank mRNA | AB209093.1 | - | 19 - 20 | 3141 | 64 |
AK293953.1 | - | 19 - 20 | 2297 | 64 | |
AK294507.1 | - | 9 - 10 | 1019 | 64 | |
BC054503.2 | - | 19 - 20 | 2984 | 64 | |
BC088829.1 | - | 19 - 20 | 3038 | 64 | |
D84476.2 | - | 19 - 20 | 2943 | 64 | |
U67156.1 | - | 19 - 20 | 3036 | 64 |