The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005923.3 | NP_005914.1 | 25 - 26 | 3876 | 80 |
XM_017010870.1 | XP_016866359.1 | 26 - 27 | 4202 | 80 | |
XM_017010871.1 | XP_016866360.1 | 26 - 27 | 4199 | 80 | |
XM_017010872.1 | XP_016866361.1 | 26 - 27 | 4201 | 80 | |
XM_017010873.1 | XP_016866362.1 | 26 - 27 | 4197 | 80 | |
XM_017010874.1 | XP_016866363.1 | 25 - 26 | 4094 | 80 | |
XM_017010875.1 | XP_016866364.1 | 25 - 26 | 4090 | 80 | |
XM_017010876.1 | XP_016866365.1 | 24 - 25 | 3210 | 80 | |
XM_017010877.1 | XP_016866366.1 | 24 - 25 | 3447 | 80 | |
GenBank mRNA | AB209093.1 | - | 25 - 26 | 3983 | 80 |
AK293953.1 | - | 25 - 26 | 3139 | 80 | |
AK294507.1 | - | 15 - 16 | 1858 | 80 | |
BC054503.2 | - | 25 - 26 | 3823 | 80 | |
BC088829.1 | - | 25 - 26 | 3877 | 80 | |
D84476.2 | - | 25 - 26 | 3782 | 80 | |
U67156.1 | - | 25 - 26 | 3875 | 80 |