The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005923.3 | NP_005914.1 | 4 - 5 | 1163 | 104 |
XM_011535839.2 | XP_011534141.2 | 5 - 6 | 1487 | 104 | |
XM_017010870.1 | XP_016866359.1 | 5 - 6 | 1486 | 104 | |
XM_017010871.1 | XP_016866360.1 | 5 - 6 | 1486 | 104 | |
XM_017010872.1 | XP_016866361.1 | 5 - 6 | 1485 | 104 | |
XM_017010873.1 | XP_016866362.1 | 5 - 6 | 1484 | 104 | |
XM_017010874.1 | XP_016866363.1 | 5 - 6 | 1486 | 104 | |
XM_017010875.1 | XP_016866364.1 | 5 - 6 | 1485 | 104 | |
XM_017010876.1 | XP_016866365.1 | 3 - 4 | 494 | 104 | |
XM_017010877.1 | XP_016866366.1 | 3 - 4 | 731 | 104 | |
GenBank mRNA | AB209093.1 | - | 4 - 5 | 1267 | 104 |
AK293953.1 | - | 4 - 5 | 423 | 104 | |
AK310942.1 | - | 5 - 6 | 942 | 104 | |
BC054503.2 | - | 4 - 5 | 1110 | 104 | |
BC088829.1 | - | 4 - 5 | 1164 | 104 | |
D84476.2 | - | 4 - 5 | 1069 | 104 | |
U67156.1 | - | 4 - 5 | 1162 | 104 |