The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053053.3 | NP_444281.1 | 4 - 5 | 515 | 94 |
GenBank mRNA | AK290964.1 | - | 4 - 5 | 332 | 94 |
AK291922.1 | - | 4 - 5 | 455 | 94 | |
BC015401.2 | - | 4 - 5 | 340 | 94 |