The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318505.1 | NP_001305434.1 | 3 - 4 | 467 | 71 |
NM_014606.2 | NP_055421.1 | 3 - 4 | 467 | 71 | |
XM_005263327.3 | XP_005263384.1 | 4 - 5 | 594 | 71 | |
XM_005263329.3 | XP_005263386.1 | 3 - 4 | 467 | 71 | |
XM_005263330.3 | XP_005263387.1 | 3 - 4 | 467 | 71 | |
XM_005263331.1 | XP_005263388.1 | 3 - 4 | 467 | 71 | |
XM_005263332.3 | XP_005263389.1 | 3 - 4 | 467 | 71 | |
XM_017008806.1 | XP_016864295.1 | 4 - 5 | 594 | 71 | |
GenBank mRNA | AK289990.1 | - | 3 - 4 | 410 | 71 |
BC038960.1 | - | 3 - 4 | 405 | 71 | |
D25215.1 | - | 3 - 4 | 394 | 71 | |
DA333248.1 | - | 3 - 4 | 467 | 71 |