Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
Chromosome Location
Chr.4: 88725954 - 89111398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015045.2 NP_001015045.1 11 - 12 1529 84
NM_001265578.1 NP_001252507.1 10 - 11 1487 84
NM_001265579.1 NP_001252508.1 11 - 12 1529 84
NM_001265580.1 NP_001252509.1 9 - 10 1445 84
NM_014883.3 NP_055698.2 17 - 18 2381 84
XM_005262681.2 XP_005262738.1 16 - 17 2339 84
XM_005262682.2 XP_005262739.1 17 - 18 2480 84
XM_005262683.2 XP_005262740.1 17 - 18 2381 84
XM_005262684.3 XP_005262741.1 16 - 17 2160 84
XM_006714057.3 XP_006714120.1 14 - 15 1656 84
XM_011531516.1 XP_011529818.1 18 - 19 2402 84
XM_011531517.1 XP_011529819.1 16 - 17 2297 84
XM_011531518.1 XP_011529820.1 18 - 19 2531 84
XM_011531519.2 XP_011529821.1 16 - 17 2090 84
XM_017007624.1 XP_016863113.1 16 - 17 2339 84
XM_017007625.1 XP_016863114.1 16 - 17 2000 84
XM_017007626.1 XP_016863115.1 14 - 15 1634 84
XM_017007627.1 XP_016863116.1 16 - 17 2225 84
XM_017007628.1 XP_016863117.1 15 - 16 2118 84
XM_017007629.1 XP_016863118.1 17 - 18 4112 84
XM_017007630.1 XP_016863119.1 15 - 16 2025 84
XM_017007631.1 XP_016863120.1 16 - 17 4070 84
XM_017007632.1 XP_016863121.1 14 - 15 1964 84
XM_017007633.1 XP_016863122.1 16 - 17 4028 84
XM_017007634.1 XP_016863123.1 10 - 11 1438 84
GenBank mRNA AB020721.2 - 11 - 12 1246 84
AK027138.1 - 11 - 12 1369 84
AK091328.1 - 4 - 5 1140 84
AK091787.1 - 10 - 11 1647 84
AK296932.1 - 9 - 10 1314 84
BC086875.1 - 10 - 11 1328 84

View Full Product Details