Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
Chromosome Location
Chr.4: 88725954 - 89111398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015045.2 NP_001015045.1 17 - 18 2328 119
NM_001265578.1 NP_001252507.1 16 - 17 2286 119
NM_001265579.1 NP_001252508.1 16 - 17 2244 119
NM_001265580.1 NP_001252509.1 15 - 16 2244 119
NM_014883.3 NP_055698.2 23 - 24 3180 119
XM_005262681.2 XP_005262738.1 22 - 23 3138 119
XM_005262682.2 XP_005262739.1 23 - 24 3279 119
XM_005262683.2 XP_005262740.1 22 - 23 3096 119
XM_005262684.3 XP_005262741.1 22 - 23 2959 119
XM_006714057.3 XP_006714120.1 20 - 21 2455 119
XM_011531516.1 XP_011529818.1 24 - 25 3201 119
XM_011531517.1 XP_011529819.1 22 - 23 3096 119
XM_011531518.1 XP_011529820.1 24 - 25 3330 119
XM_011531519.2 XP_011529821.1 22 - 23 2889 119
XM_017007624.1 XP_016863113.1 21 - 22 3054 119
XM_017007625.1 XP_016863114.1 22 - 23 2799 119
XM_017007626.1 XP_016863115.1 20 - 21 2433 119
XM_017007627.1 XP_016863116.1 22 - 23 3024 119
XM_017007628.1 XP_016863117.1 20 - 21 2833 119
XM_017007629.1 XP_016863118.1 23 - 24 4911 119
XM_017007630.1 XP_016863119.1 21 - 22 2824 119
XM_017007631.1 XP_016863120.1 22 - 23 4869 119
XM_017007632.1 XP_016863121.1 20 - 21 2763 119
XM_017007633.1 XP_016863122.1 22 - 23 4827 119
XM_017007634.1 XP_016863123.1 15 - 16 2153 119
GenBank mRNA AB020721.2 - 16 - 17 1961 119
AK027138.1 - 17 - 18 2168 119
AK091328.1 - 9 - 10 1855 119
AK123724.1 - 1 - 2 699 119
AK296932.1 - 15 - 16 2113 119
AK298262.1 - 3 - 4 401 119
BC086875.1 - 16 - 17 2127 119

View Full Product Details