The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014950.2 | NP_055765.2 | 2 - 3 | 2284 | 67 |
XM_011536568.2 | XP_011534870.1 | 4 - 5 | 2420 | 67 | |
GenBank mRNA | AB023214.1 | - | 2 - 3 | 2155 | 67 |
AY598321.1 | - | 1 - 2 | 1893 | 67 |