The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015175.2 | NP_055990.1 | 48 - 49 | 7683 | 63 |
XM_006713072.3 | XP_006713135.1 | 47 - 48 | 7599 | 63 | |
XM_011533533.2 | XP_011531835.1 | 47 - 48 | 7590 | 63 | |
XM_017006010.1 | XP_016861499.1 | 48 - 49 | 7680 | 63 | |
XM_017006011.1 | XP_016861500.1 | 48 - 49 | 7536 | 63 | |
XM_017006012.1 | XP_016861501.1 | 47 - 48 | 7599 | 63 | |
XM_017006013.1 | XP_016861502.1 | 47 - 48 | 7590 | 63 | |
XM_017006014.1 | XP_016861503.1 | 47 - 48 | 7455 | 63 | |
XM_017006015.1 | XP_016861504.1 | 46 - 47 | 7311 | 63 | |
XM_017006016.1 | XP_016861505.1 | 43 - 44 | 8257 | 63 | |
XM_017006017.1 | XP_016861506.1 | 25 - 26 | 4099 | 63 | |
GenBank mRNA | AB011112.3 | - | 34 - 35 | 5367 | 63 |
AK131104.1 | - | 48 - 49 | 7523 | 63 | |
AK293387.1 | - | 9 - 10 | 1054 | 63 | |
AK301269.1 | - | 13 - 14 | 1746 | 63 | |
AY358455.1 | - | 17 - 18 | 2543 | 63 | |
BC029944.1 | - | 10 - 11 | 1104 | 63 | |
BC060874.1 | - | 16 - 17 | 2161 | 63 |