The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145076.2 | NP_001138548.1 | 14 - 15 | 1866 | 123 |
NM_019063.4 | NP_061936.2 | 15 - 16 | 2040 | 123 | |
XM_005264267.1 | XP_005264324.1 | 16 - 17 | 2053 | 123 | |
XM_005264268.3 | XP_005264325.1 | 16 - 17 | 2053 | 123 | |
XM_006711991.1 | XP_006712054.1 | 15 - 16 | 1879 | 123 | |
XM_006711992.2 | XP_006712055.1 | 17 - 18 | 2174 | 123 | |
GenBank mRNA | AF177377.2 | - | 15 - 16 | 2002 | 123 |
AK056880.1 | - | 4 - 5 | 542 | 123 | |
AK295778.1 | - | 2 - 3 | 265 | 123 | |
AK314702.1 | - | 15 - 16 | 2014 | 123 | |
AL832392.1 | - | 2 - 3 | 261 | 123 | |
BC140845.1 | - | 15 - 16 | 1963 | 123 | |
BC146799.1 | - | 14 - 15 | 1840 | 123 | |
BX647731.1 | - | 6 - 7 | 1103 | 123 |