The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145076.2 | NP_001138548.1 | 5 - 6 | 768 | 77 |
NM_019063.4 | NP_061936.2 | 6 - 7 | 942 | 77 | |
GenBank mRNA | AF177377.2 | - | 6 - 7 | 904 | 77 |
AK314702.1 | - | 6 - 7 | 916 | 77 | |
BC140845.1 | - | 6 - 7 | 865 | 77 | |
BC146799.1 | - | 5 - 6 | 742 | 77 |