The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033.4 | NP_001024.1 | 9 - 10 | 1204 | 96 |
NM_001318064.1 | NP_001304993.1 | 8 - 9 | 1026 | 96 | |
XM_011520278.1 | XP_011518580.1 | 3 - 4 | 504 | 96 | |
GenBank mRNA | AK222786.1 | - | 9 - 10 | 1084 | 96 |
AK297988.1 | - | 9 - 10 | 949 | 96 | |
AK299629.1 | - | 7 - 8 | 841 | 96 | |
AK301891.1 | - | 3 - 4 | 344 | 96 | |
AK303394.1 | - | 8 - 9 | 870 | 96 | |
AK311593.1 | - | 3 - 4 | 509 | 96 | |
BC006498.2 | - | 9 - 10 | 1035 | 96 | |
X59543.1 | - | 9 - 10 | 1063 | 96 | |
X59617.1 | - | 9 - 10 | 1057 | 96 |