Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRAK2
Entrez Gene ID
66008
Gene Name
trafficking kinesin protein 2
Gene Aliases
ALS2CR3, CALS-C, GRIF-1, GRIF1, MILT2, OIP98
Chromosome Location
Chr.2: 201377207 - 201451596 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015049.2 NP_055864.2 11 - 12 1645 81
XM_011511690.1 XP_011509992.1 11 - 12 1489 81
XM_017004772.1 XP_016860261.1 10 - 11 1549 81
GenBank mRNA AB038951.1 - 11 - 12 1580 81
AK314384.1 - 11 - 12 1505 81
AL832573.2 - 11 - 12 1662 81
BC040668.2 - 11 - 12 1596 81
BC041330.2 - 11 - 12 1596 81
BC048093.1 - 11 - 12 1596 81
CA396280.1 - 3 - 4 285 81
DA575492.1 - 4 - 5 400 81

View Full Product Details