The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020387.3 | NP_065120.2 | 3 - 4 | 684 | 71 |
NR_133653.1 | 2 - 3 | 488 | 71 | ||
GenBank mRNA | AF083124.1 | - | 3 - 4 | 645 | 71 |
AF274025.1 | - | 3 - 4 | 663 | 71 | |
BC004416.1 | - | 1 - 2 | 174 | 71 | |
BC009831.2 | - | 3 - 4 | 658 | 71 | |
BC033322.1 | - | 3 - 4 | 648 | 71 | |
BM981225.1 | - | 2 - 3 | 71 | ||
BP273992.1 | - | 2 - 3 | 473 | 71 |