Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO30
Entrez Gene ID
84085
Gene Name
F-box protein 30
Gene Aliases
Fbx30
Chromosome Location
Chr.6: 145798135 - 145864409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032145.4 NP_115521.3 1 - 2 182 80
GenBank mRNA BC024326.1 - 1 - 2 150 80
CD109841.1 - 1 - 2 215 80

View Full Product Details