The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184745.1 | NP_001171674.1 | 3 - 4 | 223 | 64 |
NM_033215.4 | NP_149992.3 | 3 - 4 | 1166 | 64 | |
XM_005272687.3 | XP_005272744.1 | 3 - 4 | 1184 | 64 | |
XM_017029932.1 | XP_016885421.1 | 3 - 4 | 5594 | 64 | |
XM_017029933.1 | XP_016885422.1 | 3 - 4 | 5591 | 64 | |
GenBank mRNA | AK056909.1 | - | 3 - 4 | 64 | |
AK303738.1 | - | 4 - 5 | 489 | 64 | |
BC131588.1 | - | 3 - 4 | 196 | 64 | |
DC412626.1 | - | 4 - 5 | 491 | 64 |