Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CRISP2
- Entrez Gene ID
-
7180
- Gene Name
- cysteine rich secretory protein 2
- Gene Aliases
-
CRISP-2, CT36, GAPDL5, TPX1, TSP1
- Chromosome Location
-
Chr.6: 49676665 - 49714198 on Build GRCh38
View Full Product Details