The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032259.3 | NP_115635.1 | 3 - 4 | 2119 | 68 |
XM_011522699.2 | XP_011521001.1 | 4 - 5 | 2623 | 68 | |
GenBank mRNA | AL136863.1 | - | 3 - 4 | 2096 | 68 |
BC008025.1 | - | 3 - 4 | 1584 | 68 | |
BC009761.2 | - | 3 - 4 | 1119 | 68 |