Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIWIL1
Entrez Gene ID
9271
Gene Name
piwi like RNA-mediated gene silencing 1
Gene Aliases
CT80.1, HIWI, MIWI, PIWI
Chromosome Location
Chr.12: 130337887 - 130412708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190971.1 NP_001177900.1 11 - 12 1562 72
NM_004764.4 NP_004755.2 11 - 12 1562 72
XM_011539002.2 XP_011537304.1 11 - 12 1436 72
XM_011539003.2 XP_011537305.1 11 - 12 1367 72
XM_011539004.2 XP_011537306.1 11 - 12 1505 72
XM_011539005.1 XP_011537307.1 11 - 12 1470 72
XM_011539006.2 XP_011537308.1 3 - 4 362 72
XM_017020229.1 XP_016875718.1 11 - 12 1470 72
GenBank mRNA AB274731.1 - 10 - 11 1291 72
AF104260.2 - 11 - 12 1347 72
AF264004.1 - 8 - 9 1033 72
AF387507.1 - 11 - 12 1354 72
AK093133.1 - 11 - 12 1344 72
BC028581.2 - 11 - 12 1359 72

View Full Product Details