The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190971.1 | NP_001177900.1 | 2 - 3 | 353 | 75 |
NM_004764.4 | NP_004755.2 | 2 - 3 | 353 | 75 | |
XM_011539002.2 | XP_011537304.1 | 2 - 3 | 227 | 75 | |
XM_011539003.2 | XP_011537305.1 | 2 - 3 | 158 | 75 | |
XM_011539004.2 | XP_011537306.1 | 2 - 3 | 296 | 75 | |
XM_011539005.1 | XP_011537307.1 | 2 - 3 | 261 | 75 | |
XM_017020229.1 | XP_016875718.1 | 2 - 3 | 261 | 75 | |
GenBank mRNA | AF104260.2 | - | 2 - 3 | 138 | 75 |
AF387507.1 | - | 2 - 3 | 145 | 75 | |
AK093133.1 | - | 2 - 3 | 135 | 75 | |
BC028581.2 | - | 2 - 3 | 150 | 75 | |
BG718140.1 | - | 2 - 3 | 190 | 75 |