Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACOX2
Entrez Gene ID
8309
Gene Name
acyl-CoA oxidase 2
Gene Aliases
BCOX, BRCACOX, BRCOX, THCCox
Chromosome Location
Chr.3: 58505136 - 58537202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003500.3 NP_003491.1 11 - 12 1699 75
XM_005265505.1 XP_005265562.1 11 - 12 1699 75
XM_006713340.3 XP_006713403.1 11 - 12 1539 75
XM_017007302.1 XP_016862791.1 10 - 11 1701 75
GenBank mRNA AK298402.1 - 9 - 10 1253 75
AK313512.1 - 11 - 12 1644 75
BC047700.1 - 11 - 12 1678 75
X95190.1 - 10 - 11 1616 75

View Full Product Details