The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003500.3 | NP_003491.1 | 2 - 3 | 337 | 112 |
XM_005265505.1 | XP_005265562.1 | 2 - 3 | 337 | 112 | |
GenBank mRNA | AK313512.1 | - | 2 - 3 | 282 | 112 |
BC047700.1 | - | 2 - 3 | 316 | 112 | |
BG698950.1 | - | 2 - 3 | 446 | 112 | |
X95190.1 | - | 1 - 2 | 254 | 112 |