The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003500.3 | NP_003491.1 | 5 - 6 | 759 | 90 |
XM_005265505.1 | XP_005265562.1 | 5 - 6 | 759 | 90 | |
XM_006713340.3 | XP_006713403.1 | 5 - 6 | 599 | 90 | |
XM_017007302.1 | XP_016862791.1 | 4 - 5 | 761 | 90 | |
GenBank mRNA | AK313512.1 | - | 5 - 6 | 704 | 90 |
BC047700.1 | - | 5 - 6 | 738 | 90 | |
X95190.1 | - | 4 - 5 | 676 | 90 |